ENAH90822039 Enamine

(1r,3s)-3-(2-aminoethyl)cyclobutan-1-ol hydrochloride

  • MDL number MFCD28893841
  • Cas Number : 2008714-48-3
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1r,3s)-3-(2-aminoethyl)cyclobutan-1-ol hydrochloride
Inchi Code 1S/C6H13NO.ClH/c7-2-1-5-3-6(8)4-5;/h5-6,8H,1-4,7H2;1H/t5-,6+;
Inchi Key MTKMKFLGNYCBBM-KNCHESJLSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7562808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7562808
Availability details for
Quantity
  • Availability
  • Add to Cart