ENAH9082213B Enamine

3-(4-bromophenyl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD01687495

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7501667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7501667
Availability details for
Quantity
  • Availability
  • Add to Cart