ENAH9082215E Enamine

(4-bromo-2-chloropyridin-3-yl)methanol

  • MDL number MFCD20923606
  • Cas Number : 1805568-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7491827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7491827
Availability details for
Quantity
  • Availability
  • Add to Cart