ENAH90822178 Enamine

3-(1-amino-2-hydroxyethyl)phenol hydrochloride

  • MDL number MFCD28411249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7383502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7383502
Availability details for
Quantity
  • Availability
  • Add to Cart