ENAH90822180 Enamine

tert-butyl 7-hydroxy-2,5-diazaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD31743138
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7461759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7461759
Availability details for
Quantity
  • Availability
  • Add to Cart