ENAH908222D0 Enamine

(1R)-cyclohex-3-en-1-amine hydrochloride

  • MDL number MFCD24644852
  • Molecular Weight : 133.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-cyclohex-3-en-1-amine hydrochloride
Inchi Code 1S/C6H11N.ClH/c7-6-4-2-1-3-5-6;/h1-2,6H,3-5,7H2;1H/t6-;/m0./s1
Inchi Key OTEBFUBTLSFFRL-RGMNGODLSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7551555

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7551555
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart