ENAH90822315 Enamine

4-(4-fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine

  • MDL number MFCD21363743
  • Cas Number : 69675-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7509013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7509013
Availability details for
Quantity
  • Availability
  • Add to Cart