ENAH908224ED Enamine

3-(chloromethyl)-5-(1,1-difluoroethyl)pyridine hydrochloride

  • MDL number MFCD32268705
  • Cas Number : 2408963-24-4
  • Molecular Weight : 228.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7536739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7536739
Availability details for
Quantity
  • Availability
  • Add to Cart