ENAH908224EE Enamine

tert-butyl 5-(bromomethyl)-2,3-dihydro-1H-isoindole-2-carboxylate

  • MDL number MFCD16996078
  • Cas Number : 201342-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7494327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7494327
Availability details for
Quantity
  • Availability
  • Add to Cart