ENAH90822591 Enamine

ethyl 3-oxo-2-azabicyclo[2.2.2]octane-4-carboxylate

  • MDL number MFCD00102034
  • Cas Number : 21933-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7494538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7494538
Availability details for
Quantity
  • Availability
  • Add to Cart