ENAH908225BC Enamine

methyl (2R)-2-aminopent-4-enoate hydrochloride

  • MDL number MFCD17926576
  • Cas Number : 217440-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7416754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7416754
Availability details for
Quantity
  • Availability
  • Add to Cart