ENAH9082266F Enamine

5-bromo-4-fluoro-2-iodoaniline

  • MDL number MFCD30834158
  • Cas Number : 1265236-37-0
  • Molecular Weight : 315.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-fluoro-2-iodoaniline
Inchi Code 1S/C6H4BrFIN/c7-3-1-6(10)5(9)2-4(3)8/h1-2H,10H2
Inchi Key IWYBXVJNKBUYLA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7543136

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7543136
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart