ENAH908227BE Enamine

4-bromo-6-(trifluoromethyl)pyridazin-3-amine

  • MDL number MFCD29072561
  • Cas Number : 1310918-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6481742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6481742
Availability details for
Quantity
  • Availability
  • Add to Cart