ENAH908228D2 Enamine

4-{1-[(tert-butoxy)carbonyl]piperidin-4-yl}oxane-3-carboxylic acid

  • MDL number MFCD32648426
  • Cas Number : 2411221-49-1
  • Molecular Weight : 313.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434308
Availability details for
Quantity
  • Availability
  • Add to Cart