ENAH9082297B Enamine

(9H-fluoren-9-yl)methyl 3-(aminomethyl)-1,2,3,4-tetrahydroisoquinoline-2-carboxylate hydrochloride

  • MDL number MFCD04114885
  • Cas Number : 1187932-19-9
  • Molecular Weight : 420.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7491878

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7491878
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart