ENAH9082299A Enamine

(2E)-3-{1-[(tert-butoxy)carbonyl]pyrrolidin-2-yl}prop-2-enoic acid

  • MDL number MFCD20232529
  • Cas Number : 741269-02-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-[1-(tert-butoxycarbonyl)-2-pyrrolidinyl]-2-propenoic acid
Inchi Code 1S/C12H19NO4/c1-12(2,3)17-11(16)13-8-4-5-9(13)6-7-10(14)15/h6-7,9H,4-5,8H2,1-3H3,(H,14,15)/b7-6+
Inchi Key WVZOWSAYQHINTR-VOTSOKGWSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7544450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7544450
Availability details for
Quantity
  • Availability
  • Add to Cart