ENAH90822A1C Enamine

tert-butyl 3-(aminomethyl)-3-fluorocyclobutane-1-carboxylate

  • MDL number MFCD32268811
  • Cas Number : 2408971-46-8
  • Molecular Weight : 203.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7537484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7537484
Availability details for
Quantity
  • Availability
  • Add to Cart