ENAH90822A3A Enamine

4-bromo-2,3-dihydropyridazin-3-one

  • MDL number MFCD22491586
  • Cas Number : 22626-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-200082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200082
Availability details for
Quantity
  • Availability
  • Add to Cart