ENAH90822BD4 Enamine

5-chloro-4-methylthiophene-2-carboxylic acid

  • MDL number MFCD01830083
  • Cas Number : 74598-03-1
  • Molecular Weight : 176.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-245984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-245984
Availability details for
Quantity
  • Availability
  • Add to Cart