ENAH90822BE1 Enamine

4-acetamido-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD24391930
  • Cas Number : 1416404-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4324205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4324205
Availability details for
Quantity
  • Availability
  • Add to Cart