ENAH90822C18 Enamine

methyl 4-chloroquinoline-6-carboxylate

  • MDL number MFCD20488901
  • Cas Number : 648449-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7406850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7406850
Availability details for
Quantity
  • Availability
  • Add to Cart