ENAH90822F21 Enamine

tert-butyl (2S)-2-(N-hydroxyamino)-3-phenylpropanoate

  • MDL number MFCD31706072
  • Cas Number : 1137148-42-5
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl hydroxy-L-phenylalaninate
Inchi Code 1S/C13H19NO3/c1-13(2,3)17-12(15)11(14-16)9-10-7-5-4-6-8-10/h4-8,11,14,16H,9H2,1-3H3/t11-/m0/s1
Inchi Key VHKCSSWTIDGQIS-NSHDSACASA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7546626

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7546626
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart