ENAH90823039 Enamine

(1R,2S,9S)-7,11-diazatricyclo[7.3.1.0,2,7]tridecan-6-one

  • MDL number MFCD31540890
  • Cas Number : 18161-94-9
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R,5S,11aS)-decahydro-8H-1,5-methanopyrido[1,2-a][1,5]diazocin-8-one
Inchi Code 1S/C11H18N2O/c14-11-3-1-2-10-9-4-8(5-12-6-9)7-13(10)11/h8-10,12H,1-7H2/t8-,9+,10-/m0/s1
Inchi Key KWVYCGMBGRYVQH-AEJSXWLSSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7541904

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7541904
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart