ENAH90823097 Enamine

6-tert-butyl 2-methyl 5,6,7,8-tetrahydro-1,6-naphthyridine-2,6-dicarboxylate

  • MDL number MFCD09038018
  • Cas Number : 259809-47-7
  • Molecular Weight : 292.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7547114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7547114
Availability details for
Quantity
  • Availability
  • Add to Cart