ENAH908230BB Enamine

3-(methoxymethyl)oxetan-3-amine

  • MDL number MFCD28629620
  • Cas Number : 1554588-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-3104563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3104563
Availability details for
Quantity
  • Availability
  • Add to Cart