ENAH90823135 Enamine

4-methyl-3-nitropyridine-2-carbonitrile

  • MDL number MFCD19689064
  • Cas Number : 1378825-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2504629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2504629
Availability details for
Quantity
  • Availability
  • Add to Cart