ENAH90823149 Enamine

4,4-difluorocyclohexane-1-carbonitrile

  • MDL number MFCD11847832
  • Cas Number : 922728-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1589203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1589203
Availability details for
Quantity
  • Availability
  • Add to Cart