ENAH9082345B Enamine

1-fluoro-2,3-dimethoxybenzene

  • MDL number MFCD03411451
  • Cas Number : 394-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115402
Availability details for
Quantity
  • Availability
  • Add to Cart