ENAH9082354A Enamine

2-methanesulfonylpyrimidin-5-amine

  • MDL number MFCD08234879
  • Cas Number : 56621-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2966575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2966575
Availability details for
Quantity
  • Availability
  • Add to Cart