ENAH90823737 Enamine

3,3-dimethoxypyrrolidine hydrochloride

  • MDL number MFCD17676887
  • Cas Number : 1263283-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7420586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7420586
Availability details for
Quantity
  • Availability
  • Add to Cart