ENAH93E74050 Enamine

tert-butyl 2-fluoro-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD22580393
  • Cas Number : 1404196-40-2
  • Molecular Weight : 245.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7416957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7416957
Availability details for
Quantity
  • Availability
  • Add to Cart