ENAH93E74180 Enamine

2-(pyridin-3-yl)acetamide

  • MDL number MFCD08236789
  • Cas Number : 3724-16-1
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 121-123
Iupac Name 2-(3-pyridinyl)acetamide
Inchi Code 1S/C7H8N2O/c8-7(10)4-6-2-1-3-9-5-6/h1-3,5H,4H2,(H2,8,10)
Inchi Key YDHIMEXEGOCNHU-UHFFFAOYSA-N

Description

Melting Point 121-123
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-95711

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-95711
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart