ENAH93E74186 Enamine

{2-[(tert-butyldimethylsilyl)oxy]phenyl}methanol

  • MDL number MFCD30581956
  • Cas Number : 96013-79-5
  • Molecular Weight : 238.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-22159162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22159162
Availability details for
Quantity
  • Availability
  • Add to Cart