ENAH93E74199 Enamine

4-(trifluoromethyl)-1,3-thiazole-2-carboximidamide hydrochloride

  • MDL number MFCD21605322
  • Cas Number : 1251166-15-0
  • Molecular Weight : 231.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(trifluoromethyl)-1,3-thiazole-2-carboximidamide hydrochloride
Inchi Code 1S/C5H4F3N3S.ClH/c6-5(7,8)2-1-12-4(11-2)3(9)10;/h1H,(H3,9,10);1H
Inchi Key SRJWJSDIRPGAJK-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7505552

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7505552
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart