ENAH93E741D5 Enamine

7-bromo-2,3,4,5-tetrahydro-1-benzoxepin-5-ol

  • MDL number MFCD18373967
  • Cas Number : 1261999-53-4
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-2,3,4,5-tetrahydro-1-benzoxepin-5-ol
Inchi Code 1S/C10H11BrO2/c11-7-3-4-10-8(6-7)9(12)2-1-5-13-10/h3-4,6,9,12H,1-2,5H2
Inchi Key QGEPJFQBBYSBQE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-8149585

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-8149585
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart