ENAH93E74692 Enamine

4-bromo-1,2,3,5,6,7-hexahydro-s-indacene

  • MDL number MFCD31693027
  • Cas Number : 108722-46-9
  • Molecular Weight : 237.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 39-40
Iupac Name 4-bromo-1,2,3,5,6,7-hexahydro-s-indacene
Inchi Code 1S/C12H13Br/c13-12-10-5-1-3-8(10)7-9-4-2-6-11(9)12/h7H,1-6H2
Inchi Key JXUDTIMNBBRXGB-UHFFFAOYSA-N

Description

Melting Point 39-40
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7563716

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7563716
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart