ENAH93E7482D Enamine

9-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)bicyclo[5.2.0]nonane-8-carboxylic acid

  • Cas Number : 2418719-72-7
  • Molecular Weight : 405.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26677640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26677640
Availability details for
Quantity
  • Availability
  • Add to Cart