ENAH93E748BF Enamine

2-(4-bromothiophen-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD09701225
  • Molecular Weight : 242.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-bromo-2-thienyl)ethanamine hydrochloride
Inchi Code 1S/C6H8BrNS.ClH/c7-5-3-6(1-2-8)9-4-5;/h3-4H,1-2,8H2;1H
Inchi Key QAOJGFACAPVIKY-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7654161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7654161
Availability details for
Quantity
  • Availability
  • Add to Cart