ENAH93E748C0 Enamine

1-bromo-3-fluoro-2-iodo-5-methylbenzene

  • MDL number MFCD09800758
  • Cas Number : 1000576-09-9
  • Molecular Weight : 314.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-fluoro-2-iodo-5-methylbenzene
Inchi Code 1S/C7H5BrFI/c1-4-2-5(8)7(10)6(9)3-4/h2-3H,1H3
Inchi Key ZAPXFWWYNQUHFD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7654771

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7654771
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart