ENAH93E748D0 Enamine

4-bromo-3-(methoxycarbonyl)benzoic acid

  • MDL number MFCD22490651
  • Cas Number : 41684-08-6
  • Molecular Weight : 259.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-(methoxycarbonyl)benzoic acid
Inchi Code 1S/C9H7BrO4/c1-14-9(13)6-4-5(8(11)12)2-3-7(6)10/h2-4H,1H3,(H,11,12)
Inchi Key JZKLLJIVSIUAJD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-8815016

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-8815016
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart