ENAH93E7490A Enamine

4-chloro-6-methoxy-5-methylpyrimidine

  • MDL number MFCD13176588
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-6-methoxy-5-methylpyrimidine
Inchi Code 1S/C6H7ClN2O/c1-4-5(7)8-3-9-6(4)10-2/h3H,1-2H3
Inchi Key QJLIOQJTHRFATQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96033
Availability details for
Quantity
  • Availability
  • Add to Cart