ENAH93E74A3C Enamine

8-oxooctanoic acid

  • MDL number MFCD20638239
  • Cas Number : 929-48-6
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 43-44
Iupac Name 8-oxooctanoic acid
Inchi Code 1S/C8H14O3/c9-7-5-3-1-2-4-6-8(10)11/h7H,1-6H2,(H,10,11)
Inchi Key UEXYJHLCLUYOFA-UHFFFAOYSA-N

Description

Melting Point 43-44
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7706761

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7706761
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart