ENAH93E74A3F Enamine

(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(1H-1,2,3,4-tetrazol-5-yl)butanoic acid

  • MDL number MFCD26407302
  • Cas Number : 954147-36-5
  • Molecular Weight : 393.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7390585

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7390585
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart