ENAH93E74A4A Enamine

methyl 5-amino-2-bromopyridine-3-carboxylate

  • MDL number MFCD28715464
  • Cas Number : 1805616-77-6
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2932836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2932836
Availability details for
Quantity
  • Availability
  • Add to Cart