ENAH93E74ABA Enamine

(2R)-2-{[(tert-butoxy)carbonyl]amino}-4,4-dimethylpentanoic acid

  • MDL number MFCD00798621
  • Cas Number : 112695-98-4
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6472514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6472514
Availability details for
Quantity
  • Availability
  • Add to Cart