ENAH93E74B24 Enamine

tert-butyl N-[3-(aminomethyl)oxolan-3-yl]carbamate

  • MDL number MFCD12408618
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(aminomethyl)tetrahydro-3-furanylcarbamate
Inchi Code 1S/C10H20N2O3/c1-9(2,3)15-8(13)12-10(6-11)4-5-14-7-10/h4-7,11H2,1-3H3,(H,12,13)
Inchi Key GBPPASYXYUJDQC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75039
Availability details for
Quantity
  • Availability
  • Add to Cart