ENAH93E74DC5 Enamine

(2R)-2-cyclobutyl-2-hydroxyacetic acid

  • MDL number MFCD31705873
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-22142231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22142231
Availability details for
Quantity
  • Availability
  • Add to Cart