ENAH93E74DFB Enamine

2-(trifluoromethoxy)pyridine-3-carboxylic acid

  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(trifluoromethoxy)nicotinic acid
Inchi Code 1S/C7H4F3NO3/c8-7(9,10)14-5-4(6(12)13)2-1-3-11-5/h1-3H,(H,12,13)
Inchi Key XUHDHRKJBUCYSC-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-8776414
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart