ENAH93E74DFC Enamine

1-[(tert-butoxy)carbonyl]-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD22681811
  • Cas Number : 1252855-23-4
  • Molecular Weight : 212.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butoxycarbonyl)-1H-pyrazole-3-carboxylic acid
Inchi Code 1S/C9H12N2O4/c1-9(2,3)15-8(14)11-5-4-6(10-11)7(12)13/h4-5H,1-3H3,(H,12,13)
Inchi Key LBNMIKIVJHHDNF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-9601192

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-9601192
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart