ENAH93E74EAF Enamine

tert-butyl (2R)-2-methyl-3-oxoazetidine-1-carboxylate

  • MDL number MFCD28501894
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (R)-2-methyl-3-oxoazetidine-1-carboxylate
Inchi Code 1S/C9H15NO3/c1-6-7(11)5-10(6)8(12)13-9(2,3)4/h6H,5H2,1-4H3/t6-/m1/s1
Inchi Key OPQMXTJLDQEDTI-ZCFIWIBFSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7541620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7541620
Availability details for
Quantity
  • Availability
  • Add to Cart